5-Chloro-o-cresol 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
1-Chloro-3-hydroxy-4-methylbenzene, 5-Chloro-2-methylphenol
B25527-06
B25527-14
CAAAAB25527-03EA
139.32
CAD
CAAAAB25527-03
CAAAAB25527-06
CAAAAB25527-14
5-Chloro-o-cresol 98%
5-Chloro-o-cresol
Purity: 98%
CAS No.: 5306-98-9
Molecular Formula: C7H7ClO
Formula Weight: 142.59
Melting Point: 70-75°
Boiling Point: 109-112°/15mm
UN No.: 3437
MDL No.: MFCD00060672
Formula:
C₇H₇ClO MW: 142.58 g/mol Boiling Pt: 225 Melting Pt: 70…75 °C Storage Temperature: Ambient |
MDL Number:
MFCD00060672 CAS Number: 5306-98-9 EINECS: 226-160-3 UN: 3437 ADR: 6.1 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...