Indole-5-carbaldehyd 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Indole-5-carboxaldehyde, 1H-Indole-5-carboxaldehyde

B25517-03 B25517-06
CAAAAB25517-03EA 68.57 CAD
CAAAAB25517-03 CAAAAB25517-06
Indole-5-carbaldehyd 98%
Indole-5-carbaldehyd

Purity: 98% CAS No.: 1196-69-6 Molecular Formula: C9H7NO Formula Weight: 145.16 Melting Point: 96-100° MDL No.: MFCD02093664

Formula: C₉H₇NO
MW: 145.16 g/mol
Melting Pt: 96…100 °C
Storage Temperature: Ambient
MDL Number: MFCD02093664
CAS Number: 1196-69-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR