(±)-2-Phenyl-3-butyn-2-ol 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B25515-06
B25515-14
B25515-22
CAAAAB25515-06EA
75.37
CAD
CAAAAB25515-06
CAAAAB25515-14
CAAAAB25515-22
(±)-2-Phenyl-3-butyn-2-ol 98%
(±)-2-Phenyl-3-butyn-2-ol
Purity: 98%
CAS No.: 127-66-2
Molecular Formula: C10H10O
Formula Weight: 146.19
Flash Point: 96°(205°F)
Melting Point: 49-53°
Boiling Point: 92-93°/10mm
Beilstein Reference: 1100096
MDL No.: MFCD00004454
Formula:
C₁₀H₁₀O Boiling Pt: 92…93 °C (10 mmHg) Melting Pt: 49…53 °C Density: 1,031 Flash Pt: 96 °C (205 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004454 CAS Number: 127-66-2 EINECS: 204-855-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...