1-Bromo-3,5-dichlorobenzene 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B25492-06
B25492-14
B25492-22
CAAAAB25492-06EA
37.29
CAD
CAAAAB25492-06
CAAAAB25492-14
CAAAAB25492-22
1-Bromo-3,5-dichlorobenzene 97%
1-Bromo-3,5-dichlorobenzene
Purity: 97%
CAS No.: 19752-55-7
Molecular Formula: C6H3BrCl2
Formula Weight: 225.91
Melting Point: 73-77°
Boiling Point: 232°
MDL No.: MFCD00000584
Formula:
C₆H₃BrCl₂ Boiling Pt: 232 °C Melting Pt: 73…77 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000584 CAS Number: 19752-55-7 EINECS: 243-270-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...