4-Iodocumene 97%

Supplier: Thermo Scientific Chemicals
Warning

B25476-06 B25476-14
CAAAAB25476-06EA 84.06 CAD
CAAAAB25476-06 CAAAAB25476-14
4-Iodocumene 97%
4-Iodocumene

Purity: 97% CAS No.: 17356-09-1 Molecular Formula: C9H11I Formula Weight: 246.09 Refractive Index: 1.5780 Boiling Point: 236-238° MDL No.: MFCD00013711

Formula: C₉H₁₁I
Boiling Pt: 236…238 °C
Density: 1,525
Storage Temperature: Ambient
MDL Number: MFCD00013711
CAS Number: 17356-09-1
EINECS: 241-384-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR