1-Methylbenzimidazole 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-Methyl-1H-benzimidazole

B25472-06 B25472-14
CAAAAB25472-06EA 96.54 CAD
CAAAAB25472-06 CAAAAB25472-14
1-Methylbenzimidazole 99%
1-Methylbenzimidazole

Purity: 99% CAS No.: 1632-83-3 Molecular Formula: C8H8N2 Formula Weight: 132.17 Melting Point: 59-62° Boiling Point: 154°/12mm MDL No.: MFCD00192275

Formula: C₈H₈N₂
MW: 132.16 g/mol
Boiling Pt: 154 °C (12 mmHg)
Melting Pt: 59…62 °C
Storage Temperature: Ambient
MDL Number: MFCD00192275
CAS Number: 1632-83-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR