4-Iodopyridine 96%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B25459-03
B25459-06
CAAAAB25459-03EA
131.3
CAD
CAAAAB25459-03
CAAAAB25459-06
4-Iodopyridine 96%
4-Iodopyridine
Purity: 96%
CAS No.: 15854-87-2
Molecular Formula: C5H4IN
Formula Weight: 205.00
Storage Temperature: 2°C to 8°C
Melting Point: 96-99°
Beilstein Reference: 1594
MDL No.: MFCD02093937
Formula:
C₅H₄IN Melting Pt: 96…99 °C Storage Temperature: Freezer |
MDL Number:
MFCD02093937 CAS Number: 15854-87-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...