3-Formylphenylboronic acid
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Dihydroxy(3-formylphenyl)borane, 3-Boronobenzaldehyde, (3-Formylphenyl)metaboric acid, 3-Formylbenzeneboronic acid, (3-Formylphenyl)boronic acid
B25437-03
B25437-06
B25437-14
CAAAAB25437-03EA
51.03
CAD
CAAAAB25437-03
CAAAAB25437-06
CAAAAB25437-14
3-Formylphenylboronic acid
3-Formylphenylboronic acid
CAS No.: 87199-16-4
Molecular Formula: C7H7BO3
Formula Weight: 149.94
Melting Point: 109-113°
Beilstein Reference: 3030769
MDL No.: MFCD00161356
Formula:
C₇H₇BO₃ MW: 149.94 Melting Pt: 109…113 °C Storage Temperature: Ambient |
MDL Number:
MFCD00161356 CAS Number: 87199-16-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...