2-Formylphenylboronic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2-Formylbenzeneboronic acid, (2-Formylphenyl)metaboric acid, 2-Boronobenzaldehyde, (2-Formylphenyl)boronic acid, Dihydroxy(2-formylphenyl)borane
B25434-03
B25434-06
CAAAAB25434-03EA
49.77
CAD
CAAAAB25434-03
CAAAAB25434-06
2-Formylphenylboronic acid 97%
2-Formylphenylboronic acid
Purity: 97%
CAS No.: 40138-16-7
Molecular Formula: C7H7BO3
Formula Weight: 149.94
Melting Point: ca 115-120°
Beilstein Reference: 3030776
Formula:
C₇H₇BO₃ MW: 149.94 g/mol Melting Pt: ∼ 115…120 °C Storage Temperature: Ambient |
MDL Number:
MFCD00151822 CAS Number: 40138-16-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...