2-Amino-5-nitrobenzonitrile 95%

Supplier: Thermo Scientific Chemicals
Warning

B25416-14 B25416-22 B25416-36
CAAAAB25416-14EA 58.14 CAD
CAAAAB25416-14 CAAAAB25416-22 CAAAAB25416-36
2-Amino-5-nitrobenzonitrile 95%
2-Amino-5-nitrobenzonitrile

Purity: 95% CAS No.: 17420-30-3 Molecular Formula: C7H5N3O2 Formula Weight: 163.14 Melting Point: 207-210° MDL No.: MFCD00007362

Formula: C₇H₅N₃O₂
Melting Pt: 207…210 °C
Storage Temperature: Ambient
MDL Number: MFCD00007362
CAS Number: 17420-30-3
EINECS: 241-446-8
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR