Bis(4-methoxyphenyl) disulfide 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B25399-03
B25399-06
CAAAAB25399-03EA
101.59
CAD
CAAAAB25399-03
CAAAAB25399-06
Bis(4-methoxyphenyl) disulfide 97%
Bis(4-methoxyphenyl) disulphide
Purity: 97%
CAS No.: 5335-87-5
Molecular Formula: C14H14O2S2
Formula Weight: 278.39
Melting Point: 37-41°
Boiling Point: 200°/3mm
MDL No.: MFCD00041358
Formula:
C₁₄H₁₄O₂S₂ Boiling Pt: 200 °C/3 mm Melting Pt: 37…41 °C Storage Temperature: Ambient |
MDL Number:
MFCD00041358 CAS Number: 5335-87-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...