ω,3,4-Trichloroacetophenone
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
α,3,4-Trichloroacetophenone, 2-Chloro-1-(3,4-dichlorophenyl)ethanone, 3,4-Dichlorophenacyl chloride, 2,3',4'-Trichloroacetophenone
B25397-06
B25397-14
CAAAAB25397-06EA
76.47
CAD
CAAAAB25397-06
CAAAAB25397-14
ω,3,4-Trichloroacetophenone
ω,3,4-Trichloroacetophenone
CAS No.: 42981-08-8
Molecular Formula: C8H5Cl3O
Formula Weight: 223.49
Melting Point: 43-45°
Boiling Point: 171-173°/15mm
UN No.: 1759
MDL No.: MFCD03970382
Formula:
C₈H₅Cl₃O MW: 223.49 g/mol Boiling Pt: 171…173 °C/15 mm Melting Pt: 43…45 °C Storage Temperature: Ambient |
MDL Number:
MFCD03970382 CAS Number: 42981-08-8 UN: 1759 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...