2-Bromo-5-fluorobenzoic acid 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B25394-06
B25394-14
CAAAAB25394-03EA
144.27
CAD
CAAAAB25394-03
CAAAAB25394-06
CAAAAB25394-14
2-Bromo-5-fluorobenzoic acid 98+%
2-Bromo-5-fluorobenzoic acid
Purity: 98+%
CAS No.: 394-28-5
Molecular Formula: C7H4BrFO2
Formula Weight: 219.01
Melting Point: 154-158°
Beilstein Reference: 2575978
Formula:
C₇H₄BrFO₂ Melting Pt: 154…158 °C Storage Temperature: Ambient |
MDL Number:
MFCD00142874 CAS Number: 394-28-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...