4-Iodobenzoyl chloride 98%

Supplier: Thermo Scientific Chemicals
Danger

B25388-06 B25388-14 B25388-22
CAAAAB25388-06EA 119.92 CAD
CAAAAB25388-06 CAAAAB25388-14 CAAAAB25388-22
4-Iodobenzoyl chloride 98%
4-Iodobenzoyl chloride

Purity: 98% CAS No.: 1711-02-0 Molecular Formula: C7H4ClIO Formula Weight: 266.47 Flash Point: 110°(230°F) Melting Point: 61-65° Boiling Point: 120-121°/1mm Beilstein Reference: 774718 UN No.: 3261 MDL No.: MFCD00001058

Formula: C₇H₄ClIO
Boiling Pt: 120…121 °C/1 mm
Melting Pt: 61…65 °C
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001058
CAS Number: 1711-02-0
EINECS: 216-974-7
UN: 3261
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR