4-Bromo-m-toluonitrile 98%

Supplier: Thermo Scientific Chemicals
Warning

B25377-03 B25377-06 B25377-14
CAAAAB25377-03EA 41.24 CAD
CAAAAB25377-03 CAAAAB25377-06 CAAAAB25377-14
4-Bromo-m-toluonitrile 98%
4-Bromo-m-toluonitrile

Purity: 98% CAS No.: 41963-20-6 Molecular Formula: C8H6BrN Formula Weight: 196.05 Flash Point: 96°(205°F) Melting Point: 52-54° Boiling Point: 129-130°/5mm Beilstein Reference: 2354972 UN No.: 3439 MDL No.: MFCD00031538

Formula: C₈H₆BrN
MW: 196.05 g/mol
Boiling Pt: 129…130 °C/5 mm
Melting Pt: 52…54 °C
Flash Pt: 96 °C (205 °F)
Storage Temperature: Ambient
MDL Number: MFCD00031538
CAS Number: 41963-20-6
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR