2'-Iodoacetophenone 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B25345-03
B25345-06
CAAAAB25345-03EA
57.51
CAD
CAAAAB25345-03
CAAAAB25345-06
2'-Iodoacetophenone 98+%
2'-Iodoacetophenone
Purity: 98+%
CAS No.: 2142-70-3
Molecular Formula: C8H7IO
Formula Weight: 246.05
Flash Point: 103°(218°F)
Refractive Index: 1.6180
Boiling Point: 139-140°/12mm
Beilstein Reference: 2325078
MDL No.: MFCD00094998
Formula:
C₈H₇IO Boiling Pt: 139…140 °C/12 mm Density: 1.746 Flash Pt: 103 °C (218 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00094998 CAS Number: 2142-70-3 UN: 2810 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...