3,4-Dichlorocinnamic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B25308-06
B25308-14
CAAAAB25308-06EA
57.51
CAD
CAAAAB25308-06
CAAAAB25308-14
CAAAAB25308-22
3,4-Dichlorocinnamic acid 97%
3,4-Dichlorocinnamic acid
Purity: 97%
CAS No.: 1202-39-7
Molecular Formula: C9H6Cl2O2
Formula Weight: 217.05
Melting Point: 218-220°
Beilstein Reference: 1872129
Formula:
C₉H₆Cl₂O₂ Melting Pt: 218…220 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004385 CAS Number: 1202-39-7 EINECS: 214-866-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...