4-Bromo-2-methylphenyl isothiocyanate 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
B25296-06
B25296-14
CAAAAB25296-06EA
149.47
CAD
CAAAAB25296-06
CAAAAB25296-14
4-Bromo-2-methylphenyl isothiocyanate 98%
4-Bromo-2-methylphenyl isothiocyanate
Purity: 98%
CAS No.: 19241-38-4
Molecular Formula: C8H6BrNS
Formula Weight: 228.12
Melting Point: 44-46°
Boiling Point: 129-130°/3mm
UN No.: 2923
MDL No.: MFCD00041087
Formula:
C₈H₆BrNS Boiling Pt: 129…130 °C/3 mm Melting Pt: 44…46 °C Storage Temperature: Ambient |
MDL Number:
MFCD00041087 CAS Number: 19241-38-4 EINECS: 242-907-6 UN: 2923 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...