1-Bromo-4-fluoro-2-iodobenzene 98+%

Supplier: Thermo Scientific Chemicals
Warning

B25289-06 B25289-14
CAAAAB25289-03EA 86.32 CAD
CAAAAB25289-03 CAAAAB25289-06 CAAAAB25289-14
1-Bromo-4-fluoro-2-iodobenzene 98+%
1-Bromo-4-fluoro-2-iodobenzene

Purity: 98+% CAS No.: 202865-72-3 Molecular Formula: C6H3BrFI Formula Weight: 300.90 Flash Point: 110°(230°F) Refractive Index: 1.6300 Boiling Point: 235° MDL No.: MFCD00042578

Formula: C₆H₃BrFI
Boiling Pt: 235 °C
Density: 2,300
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00042578
CAS Number: 202865-72-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR