Methyl-5-nitrosalicylate 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Methyl 2-hydroxy-5-nitrobenzoate

B25278-09 B25278-18 B25278-30
CAAAAB25278-09EA 79 CAD
CAAAAB25278-09 CAAAAB25278-18 CAAAAB25278-30
Methyl-5-nitrosalicylate 98%
Methyl 5-nitrosalicylate

Purity: 98% CAS No.: 17302-46-4 Molecular Formula: C8H7NO5 Formula Weight: 197.15 Melting Point: 114-117° MDL No.: MFCD00024695

Formula: C₈H₇NO₅
MW: 197.15 g/mol
Melting Pt: 114…117 °C
Storage Temperature: Ambient
MDL Number: MFCD00024695
CAS Number: 17302-46-4
EINECS: 241-330-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR