2-Chloro-5-iodopyridine 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B25271-03
B25271-06
B25271-14
CAAAAB25271-03EA
80.74
CAD
CAAAAB25271-03
CAAAAB25271-06
CAAAAB25271-14
2-Chloro-5-iodopyridine 98%
2-Chloro-5-iodopyridine
Purity: 98%
CAS No.: 69045-79-0
Molecular Formula: C5H3ClIN
Formula Weight: 239.44
Melting Point: 95-98°
Beilstein Reference: 108889
Formula:
C₅H₃ClIN Melting Pt: 95…98 °C Storage Temperature: Ambient |
MDL Number:
MFCD01863635 CAS Number: 69045-79-0 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...