3-Nitrophthalonitrile 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B25269-06
B25269-14
CAAAAB25269-06EA
120.24
CAD
CAAAAB25269-06
CAAAAB25269-14
3-Nitrophthalonitrile 99%
3-Nitrophthalonitrile
Purity: 99%
CAS No.: 51762-67-5
Molecular Formula: C8H3N3O2
Formula Weight: 173.13
Melting Point: 162-165°
Beilstein Reference: 2263686
Formula:
C₈H₃N₃O₂ Melting Pt: 162…165 °C Storage Temperature: Ambient |
MDL Number:
MFCD00191558 CAS Number: 51762-67-5 ADR: 6.1,I |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...