5-Bromo-2-hydroxypyridine 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
5-Bromo-2-pyridinol, 3-Bromo-6-hydroxypyridine, 3-Bromo-6-pyridinol, 5-Bromopyridin-2-ol
B25267-06
B25267-14
CAAAAB25267-06EA
41.71
CAD
CAAAAB25267-06
CAAAAB25267-14
5-Bromo-2-hydroxypyridine 97%
5-Bromo-2-hydroxypyridine
Purity: 97%
CAS No.: 13466-38-1
Molecular Formula: C5H4BrNO
Formula Weight: 174.00
Melting Point: 178-182°
Beilstein Reference: 108751
Formula:
C₅H₄BrNO MW: 174 g/mol Melting Pt: 178…182 °C Storage Temperature: Ambient |
MDL Number:
MFCD00234042 CAS Number: 13466-38-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...