Sodium alginate, low viscosity
Supplier: Thermo Scientific Chemicals
B25266-22
B25266-36
CAAAAB25266-18EA
77.53
CAD
CAAAAB25266-18
CAAAAB25266-22
CAAAAB25266-30
CAAAAB25266-36
Sodium alginate, low viscosity
Sodium alginate
A gelling polysaccharide extracted from giant brown seaweed, useful in viscosity studies
CAS No.: 9005-38-3
Merck Reference: 14,241
MDL No.: MFCD00081310
Storage Temperature:
Ambient |
MDL Number:
MFCD00081310 CAS Number: 9005-38-3 Merck Index: 14,241 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...