α-Bromo-3-nitro-p-toluic acid 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 4-(Bromomethyl)-3-nitrobenzoic acid

B25255-03 B25255-06 B25255-14
CAAAAB25255-03EA 67.31 CAD
CAAAAB25255-03 CAAAAB25255-06 CAAAAB25255-14
α-Bromo-3-nitro-p-toluic acid 97%
α-Bromo-3-nitro-p-toluic acid

Purity: 97% CAS No.: 55715-03-2 Molecular Formula: C8H6BrNO4 Formula Weight: 260.05 Melting Point: 128-133° UN No.: 3261 MDL No.: MFCD00216575

Formula: C₈H₆BrNO₄
MW: 260.04 g/mol
Melting Pt: 128…133 °C
Storage Temperature: Ambient
MDL Number: MFCD00216575
CAS Number: 55715-03-2
UN: 3261
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR