2-Bromo-4-methylpyridine 96%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Bromo-4-picoline

B25253-03 B25253-06 B25253-14
CAAAAB25253-03EA 31.92 CAD
CAAAAB25253-03 CAAAAB25253-06 CAAAAB25253-14
2-Bromo-4-methylpyridine 96%
2-Bromo-4-methylpyridine

Purity: 96% CAS No.: 4926-28-7 Molecular Formula: C6H6BrN Formula Weight: 172.03 Flash Point: 110°(230°F) Refractive Index: 1.5630 Boiling Point: 87°/10mm Beilstein Reference: 107331 MDL No.: MFCD00082590

Formula: C₆H₆BrN
MW: 172.02 g/mol
Boiling Pt: 87 °C/10 mm
Melting Pt: 218…228 °C
Density: 1.545
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00082590
CAS Number: 4926-28-7

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR