3-Chloro-3-methyl-1-butyne 95%
Supplier: Thermo Scientific Chemicals
|
Danger
|
B25252-06
B25252-14
CAAAAB25252-06EA
210.14
CAD
CAAAAB25252-06
CAAAAB25252-14
3-Chloro-3-methyl-1-butyne 95%
3-Chloro-3-methyl-1-butyne
Purity: 95%
CAS No.: 1111-97-3
Molecular Formula: C5H7Cl
Formula Weight: 102.57
Storage Temperature: 2°C to 8°C
Flash Point: 4°(40°F)
Refractive Index: 1.4178
Odor: Recognizable
Melting Point: -61°
Boiling Point: 73-75°
UN No.: 1993
MDL No.: MFCD00190221
Formula:
C₅H₇Cl MW: 102.56 g/mol Boiling Pt: 73…75 °C Melting Pt: –61 °C Density: 0.913 Flash Pt: 4 °C (40 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00190221 CAS Number: 1111-97-3 EINECS: 214-188-9 UN: 1993 ADR: 3 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...