Methyl-3-(trimethylsilyl)propiolate 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B25245-03
B25245-06
CAAAAB25245-03EA
184.86
CAD
CAAAAB25245-03
CAAAAB25245-06
CAAAAB25245-14
Methyl-3-(trimethylsilyl)propiolate 98%
Methyl 3-(trimethylsilyl)propiolate
Purity: 98%
CAS No.: 42201-71-8
Molecular Formula: C7H12O2Si
Formula Weight: 156.25
Refractive Index: 1.4400
Boiling Point: 71-75/11mm
UN No.: 3272
MDL No.: MFCD04039783
Formula:
C₇H₁₂O₂Si Boiling Pt: 71…75/11 mm Storage Temperature: Ambient |
MDL Number:
MFCD04039783 CAS Number: 42201-71-8 UN: 3272 ADR: "3",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...