4-(Trifluoromethyl)benzenesulfonamide 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
α,α,α-Trifluoro-p-toluenesulfonamide, α,α,α-Trifluoro-p-toluenesulphonamide
B25243-03
B25243-14
CAAAAB25243-03EA
56.25
CAD
CAAAAB25243-03
CAAAAB25243-06
CAAAAB25243-14
4-(Trifluoromethyl)benzenesulfonamide 97%
4-(Trifluoromethyl)benzenesulphonamide
Purity: 97%
CAS No.: 830-43-3
Molecular Formula: C7H6F3NO2S
Formula Weight: 225.19
Melting Point: 178-182°
Beilstein Reference: 2695323
Formula:
C₇H₆F₃NO₂S MW: 225.19 g/mol Melting Pt: 178…182 °C Storage Temperature: Ambient |
MDL Number:
MFCD00159251 CAS Number: 830-43-3 EINECS: 212-596-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...