Ethyl tridecanoate 97%

Supplier: Thermo Scientific Chemicals

Synonyms: Tridecanoic acid ethyl ester, Tridecylic acid ethyl ester

B25242-06 B25242-14 B25242-22
CAAAAB25242-06EA 240.16 CAD
CAAAAB25242-06 CAAAAB25242-14 CAAAAB25242-22
Ethyl tridecanoate 97%
Ethyl tridecanoate

Purity: 97% CAS No.: 28267-29-0 Molecular Formula: C15H30O2 Formula Weight: 242.41 Flash Point: 110°(230°F) Refractive Index: 1.4340 Boiling Point: 121-123°/2.2mm Beilstein Reference: 1773737 MDL No.: MFCD00008978

Formula: C₁₅H₃₀O₂
MW: 242.4 g/mol
Boiling Pt: 121…123 °C (2.2 mmHg)
Density: 0.86
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008978
CAS Number: 28267-29-0
EINECS: 248-929-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR