3',5'-Dimethoxyacetophenone 97%

Supplier: Thermo Scientific Chemicals
Warning

B25224-03 B25224-06
CAAAAB25224-03EA 147.89 CAD
CAAAAB25224-03 CAAAAB25224-06 CAAAAB25224-14
3',5'-Dimethoxyacetophenone 97%
3',5'-Dimethoxyacetophenone

Purity: 97% CAS No.: 39151-19-4 Molecular Formula: C10H12O3 Formula Weight: 180.20 Flash Point: 110°(230°F) Refractive Index: 1.5430 Melting Point: 39-41° Boiling Point: 290-291° Beilstein Reference: 2045586 MDL No.: MFCD00008739

Formula: C₁₀H₁₂O₃
Boiling Pt: 290…291 °C
Melting Pt: 39…41 °C
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008739
CAS Number: 39151-19-4
EINECS: 254-322-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR