ω-Bromo-4-methylacetophenone 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
α-Bromo-4-methylacetophenone, Bromomethyl p-methylphenyl ketone, 2-Bromo-4'-methylacetophenone, 2-Bromo-1-(p-tolyl)ethanone, 4-Methylphenacyl bromide
B25217-06
B25217-14
B25217-22
CAAAAB25217-06EA
109.02
CAD
CAAAAB25217-06
CAAAAB25217-14
CAAAAB25217-22
ω-Bromo-4-methylacetophenone 98%
ω-Bromo-4-methylacetophenone
Purity: 98%
CAS No.: 619-41-0
Molecular Formula: C9H9BrO
Formula Weight: 213.08
Flash Point:
Formula:
C₉H₉BrO MW: 213.07 g/mol Boiling Pt: 105 Melting Pt: 50…54 °C Flash Pt: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000203 CAS Number: 619-41-0 EINECS: 210-595-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...