Itaconic anhydride 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Methylenesuccinic anhydride, 3-Methylenedihydrofuran-2,5-dione

B25216-14 B25216-22 B25216-36
CAAAAB25216-14EA 60.04 CAD
CAAAAB25216-14 CAAAAB25216-22 CAAAAB25216-36
Itaconic anhydride 97%
Itaconic anhydride

Purity: 97% CAS No.: 2170-03-8 Molecular Formula: C5H4O3 Formula Weight: 112.09 Melting Point: 68-72° Boiling Point: 114-115°/12mm MDL No.: MFCD00005530

Formula: C₅H₄O₃
Boiling Pt: 114…115 °C/12 mm
Melting Pt: 68…72 °C
Storage Temperature: Ambient
MDL Number: MFCD00005530
CAS Number: 2170-03-8
EINECS: 218-518-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR