3-Bromo-2-chloropyridine 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B25206-06
B25206-14
CAAAB25206-03EA
34.93
CAD
CAAAB25206-03
CAAAB25206-06
CAAAB25206-14
3-Bromo-2-chloropyridine 97%
3-Bromo-2-chloropyridine
Purity: 97%
CAS No.: 52200-48-3
Molecular Formula: C5H3BrClN
Formula Weight: 192.44
Melting Point: 52-57°
Boiling Point: 97°/10mm
MDL No.: MFCD00234007
Formula:
C₅H₃BrClN Boiling Pt: 97 °C/10 mm Melting Pt: 52…57 °C Storage Temperature: Ambient |
MDL Number:
MFCD00234007 CAS Number: 52200-48-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...