6-Nitro-2,4-xylenol 98%

Supplier: Thermo Scientific Chemicals
Warning

B25205-06 B25205-14 B25205-22
CAAAAB25205-06EA 91.64 CAD
CAAAAB25205-06 CAAAAB25205-14 CAAAAB25205-22
6-Nitro-2,4-xylenol 98%
6-Nitro-2,4-xylenol

Purity: 98% CAS No.: 14452-34-7 Molecular Formula: C8H9NO3 Formula Weight: 167.17 Melting Point: 68-71° MDL No.: MFCD00191633

Formula: C₈H₉NO₃
Melting Pt: 68…71 °C
Storage Temperature: Ambient
MDL Number: MFCD00191633
CAS Number: 14452-34-7
EINECS: 238-436-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR