4-Formylphenylboronic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B25199-03
B25199-06
B25199-14
CAAAAB25199-03EA
62.25
CAD
CAAAAB25199-03
CAAAAB25199-06
CAAAAB25199-14
4-Formylphenylboronic acid 97%
4-Formylphenylboronic acid
Purity: 97%
CAS No.: 87199-17-5
Molecular Formula: C7H7BO3
Formula Weight: 149.94
Melting Point: 260-266°
Beilstein Reference: 3030770
Formula:
C₇H₇BO₃ Melting Pt: 260…266 °C Storage Temperature: Ambient |
MDL Number:
MFCD00151823 CAS Number: 87199-17-5 EINECS: 438-670-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...