2-Methyl-2-nitropropane-1,3-diol 97%

Supplier: Thermo Scientific Chemicals

B25196-06 B25196-14
CAAAAB25196-06EA 129.09 CAD
CAAAAB25196-06 CAAAAB25196-14 CAAAAB25196-22
2-Methyl-2-nitropropane-1,3-diol 97%
2-Methyl-2-nitropropane-1,3-diol

Purity: 97% CAS No.: 77-49-6 Molecular Formula: C4H9NO4 Formula Weight: 135.12 Melting Point: 148-151° MDL No.: MFCD00024812

Formula: C₄H₉NO₄
Boiling Pt: 324 °C (1013 hPa)
Melting Pt: 148…151 °C
Storage Temperature: Ambient
MDL Number: MFCD00024812
CAS Number: 77-49-6
EINECS: 201-031-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR