6(5H)-Phenanthridinone 96%
Supplier: Thermo Scientific Chemicals
B25187-03
B25187-06
CAAAAB25187-03EA
64.94
CAD
CAAAAB25187-03
CAAAAB25187-06
6(5H)-Phenanthridinone 96%
6(5H)-Phenanthridinone
Purity: 96%
CAS No.: 1015-89-0
Molecular Formula: C13H9NO
Formula Weight: 195.22
Melting Point: 288-294°
Beilstein Reference: 140641
Formula:
C₁₃H₉NO Melting Pt: 288…294 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004988 CAS Number: 1015-89-0 EINECS: 213-804-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...