2-(Trifluoromethyl)phenylhydrazinium chloride 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
B25182-06
B25182-14
CAAAAB25182-06EA
167.48
CAD
CAAAAB25182-06
CAAAAB25182-14
CAAAAB25182-22
2-(Trifluoromethyl)phenylhydrazinium chloride 98%
2-(Trifluoromethyl)phenylhydrazinium chloride
Purity: 98%
CAS No.: 3107-34-4
Molecular Formula: C7H7F3N2.HCl
Formula Weight: 212.60
Melting Point: ca 235° dec.
Beilstein Reference: 4582303
Formula:
C₇H₈ClF₃N₂ Melting Pt: ca 235 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00102619 CAS Number: 3107-34-4 UN: 2811 ADR: "6.1",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...