2,3,5,6-Tetrafluorobenzyl alcohol 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B25162-06
B25162-14
CAAAAB25162-03EA
76.63
CAD
CAAAAB25162-03
CAAAAB25162-06
CAAAAB25162-14
2,3,5,6-Tetrafluorobenzyl alcohol 98%
2,3,5,6-Tetrafluorobenzyl alcohol
Purity: 98%
CAS No.: 4084-38-2
Molecular Formula: C7H4F4O
Formula Weight: 180.10
Odor: Recognizable
Melting Point: 34-38°
Boiling Point: 220-225°
MDL No.: MFCD00792428
Formula:
C₇H₄F₄O Boiling Pt: 220…225 °C Melting Pt: 34…38 °C Storage Temperature: Ambient |
MDL Number:
MFCD00792428 CAS Number: 4084-38-2 EINECS: 443-840-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...