2-Naphthaldehyde 98%

Supplier: Thermo Scientific Chemicals
Warning

B25157-06 B25157-14 B25157-22
CAAAAB25157-06EA 79.63 CAD
CAAAAB25157-06 CAAAAB25157-14 CAAAAB25157-22
2-Naphthaldehyde 98%
2-Naphthaldehyde

Purity: 98% CAS No.: 66-99-9 Molecular Formula: C11H8O Formula Weight: 156.18 Melting Point: 59-62° Boiling Point: 160°/19mm MDL No.: MFCD00004094

Formula: C₁₁H₈O
Boiling Pt: 160 °C/19 mm
Melting Pt: 59…62 °C
Density: 1.0775 g/cm³ (99 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004094
CAS Number: 66-99-9
EINECS: 200-640-2
UN: 3335

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR