3,5-Dimethoxybenzyl alcohol 99%

Supplier: Thermo Scientific Chemicals

Synonyms: (3,5-Dimethoxyphenyl)methanol

B25138-06 B25138-14
CAAAAB25138-03EA 98.72 CAD
CAAAAB25138-03 CAAAAB25138-06 CAAAAB25138-14
3,5-Dimethoxybenzyl alcohol 99%
3,5-Dimethoxybenzyl alcohol

Purity: 99% CAS No.: 705-76-0 Molecular Formula: C9H12O3 Formula Weight: 168.19 Flash Point: 110°(230°F) Melting Point: 46-50° Boiling Point: 183-186°/20mm Beilstein Reference: 1866305 MDL No.: MFCD00004641

Formula: C₉H₁₂O₃
MW: 168.19 g/mol
Boiling Pt: 183…186 °C/20 mm
Melting Pt: 46…50 °C
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004641
CAS Number: 705-76-0
EINECS: 211-888-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR