Potassium-D-gluconate 99%

Supplier: Thermo Scientific Chemicals

Synonyms: Potassium Gluconate, Gluconic Acid Potassium Salt, D-Gluconic acid potassium salt, Potassium (2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoate

B25135-0B B25135-18 B25135-30
CAAAAB25135-0BEA 319.16 CAD
CAAAAB25135-0B CAAAAB25135-18 CAAAAB25135-30
Potassium-D-gluconate 99%
Potassium D-gluconate

Purity: 99% CAS No.: 299-27-4 Molecular Formula: C6H11KO7 Formula Weight: 234.25 Melting Point: ca 180° dec. MDL No.: MFCD00064211

Formula: HOH₂C(CH(OH))₄COOK
Melting Pt: ∼ 180 °C (dec.)
Density: 1.73 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064211
CAS Number: 299-27-4
EINECS: 206-074-2
Merck Index: 12,07796

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR