5-Hexynoic acid 96%

Supplier: Thermo Scientific Chemicals
Danger

B25132-03 B25132-06 B25132-14
CAAAAB25132-03EA 79.63 CAD
CAAAAB25132-03 CAAAAB25132-06 CAAAAB25132-14
5-Hexynoic acid 96%
5-Hexynoic acid

Purity: 96% CAS No.: 53293-00-8 Molecular Formula: HC≡C(CH2)3CO2H Formula Weight: 112.13 Flash Point: 110°(230°F) Refractive Index: 1.4495 Boiling Point: 73-74°/0.2mm Beilstein Reference: 1743192 UN No.: 3265 MDL No.: MFCD00066346

Formula: C₆H₈O₂
Boiling Pt: 73…74 °C/0.2 mm
Melting Pt: 41 °C
Density: 0.990
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00066346
CAS Number: 53293-00-8
UN: 2922
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR