N-(6-Bromohexyl)phthalimide 97%
Supplier: Thermo Scientific Chemicals
B25128-06
B25128-14
B25128-22
CAAAB25128-06EA
164.32
CAD
CAAAB25128-06
CAAAB25128-14
CAAAB25128-22
N-(6-Bromohexyl)phthalimide 97%
N-(6-Bromohexyl)phthalimide
Purity: 97%
CAS No.: 24566-79-8
Molecular Formula: C14H16BrNO2
Formula Weight: 310.19
Odor: Odorless
Melting Point: 54-60°
Boiling Point: 186-188°/0.4mm
Beilstein Reference: 186875
MDL No.: MFCD00023098
Formula:
C₁₄H₁₆BrNO₂ Boiling Pt: 186…188 °C/0.4 mm Melting Pt: 54…60 °C Storage Temperature: Ambient |
MDL Number:
MFCD00023098 CAS Number: 24566-79-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...