(R)-(-)-1,3-Butanediol 98%
Supplier: Thermo Scientific Chemicals
B25124-03
B25124-06
CAAAAB25124-03EA
119.29
CAD
CAAAAB25124-03
CAAAAB25124-06
(R)-(-)-1,3-Butanediol 98%
(R)-(-)-1,3-Butanediol
Purity: 98%
CAS No.: 6290-03-5
Molecular Formula: C4H10O2
Formula Weight: 90.12
Flash Point: 121°(249°F)
Refractive Index: 1.4400
Odor: Weak
Boiling Point: 203-204°
Beilstein Reference: 1718944
Optical Rotation: -30° (neat)
MDL No.: MFCD00064277
Formula:
C₄H₁₀O₂ Boiling Pt: 203…204 °C Density: 1.005 Flash Pt: 121 °C (249 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00064277 CAS Number: 6290-03-5 EINECS: 228-532-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...