4-Hydroxy-3,5-dimethylbenzaldehyde 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3,5-Dimethyl-4-hydroxybenzaldehyde

B25123-03 B25123-06 B25123-14
CAAAAB25123-03EA 67.62 CAD
CAAAAB25123-03 CAAAAB25123-06 CAAAAB25123-14
4-Hydroxy-3,5-dimethylbenzaldehyde 98%
4-Hydroxy-3,5-dimethylbenzaldehyde

Purity: 98% CAS No.: 2233-18-3 Molecular Formula: C9H10O2 Formula Weight: 150.18 Melting Point: 112-116° Beilstein Reference: 1908717

Formula: C₉H₁₀O₂
MW: 150.18 g/mol
Melting Pt: 112…116 °C
Storage Temperature: Ambient
MDL Number: MFCD00006946
CAS Number: 2233-18-3
EINECS: 218-774-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR