Cyclopropyltriphenylphosphonium bromide 98%

Supplier: Thermo Scientific Chemicals
Warning

B25102-06 B25102-14 B25102-22
CAAAAB25102-06EA 100.96 CAD
CAAAAB25102-06 CAAAAB25102-14 CAAAAB25102-22
Cyclopropyltriphenylphosphonium bromide 98%
Cyclopropyltriphenylphosphonium bromide

Purity: 98% CAS No.: 14114-05-7 Molecular Formula: C21H20BrP Formula Weight: 383.27 Melting Point: 183-186° Beilstein Reference: 6413483

Formula: C₂₁H₂₀BrP
MW: 383.27 g/mol
Melting Pt: 183…186 °C
Storage Temperature: Ambient
MDL Number: MFCD00011872
CAS Number: 14114-05-7
EINECS: 237-970-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR