Trimethylsulfonium bromide 98%

Supplier: Thermo Scientific Chemicals
Warning

B25098-14 B25098-22 B25098-36
CAAAAB25098-14EA 85.64 CAD
CAAAAB25098-14 CAAAAB25098-22 CAAAAB25098-36
Trimethylsulfonium bromide 98%
Trimethylsulphonium bromide

Purity: 98% CAS No.: 3084-53-5 Molecular Formula: (CH3)3SBr Formula Weight: 157.07 Melting Point: 203° subl. MDL No.: MFCD00051367

Formula: C₃H₉BrS
Melting Pt: 203 °C (subl.)
Density: 1,67 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00051367
CAS Number: 3084-53-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR