4,4'-Dimethylbiphenyl 99%

Supplier: Thermo Scientific Chemicals

B25089-03 B25089-06 B25089-14
CAAAAB25089-03EA 66.36 CAD
CAAAAB25089-03 CAAAAB25089-06 CAAAAB25089-14
4,4'-Dimethylbiphenyl 99%
4,4'-Dimethylbiphenyl

Purity: 99% CAS No.: 613-33-2 Molecular Formula: C14H14 Formula Weight: 182.27 Melting Point: 118-122° Boiling Point: 295° MDL No.: MFCD00008545

Formula: C₁₄H₁₄
Boiling Pt: 295 °C
Melting Pt: 118…122 °C
Density: 1.102 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008545
CAS Number: 613-33-2
EINECS: 210-337-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR